Molybdenum(VI) oxide, EMSURE® for analysis, Supelco®

Supplier: MilliporeSigma
Warning

1.00403.0100 1.00403.0500
EM1.00403.0100CS 31679 USD
EM1.00403.0100 EM1.00403.0500
Molybdenum(VI) oxide, EMSURE® for analysis, Supelco®
Molybdenum (VI) oxide
Formula: MoO₃
MDL Number: MFCD00003469
CAS Number: 1313-27-5
UN: 3288
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR