N,N-Dimethylacetamide, SupraSolv® for headspace gas chromatography, Supelco®

Supplier: MilliporeSigma
Danger

1.00399.1000
EM1.00399.1000CS 3693 USD
EM1.00399.1000
N,N-Dimethylacetamide, SupraSolv® for headspace gas chromatography, Supelco®
N,N-Dimethylacetamide

SupraSolv® solvents for headspace gas chromatography are developed particularly for the analysis of residual solvents in drug substances, excipients and drug products according to ICH, Ph Eur, and USP - SupraSolv® headspace solvents are tested for all residual solvents defined by the ICH.

Formula: CH₃CON(CH₃)₂
Boiling Pt: 166.1 °C (1013 hPa)
Melting Pt: –20 °C
Density: 0.937 g/cm³ (20 °C)
Flash Pt: 70 °C
Storage Temperature: Ambient
MDL Number: MFCD00008686
CAS Number: 127-19-5
EINECS: 204-826-4
UN: 2810
ADR: 6.1,III
Merck Index: 13,03254
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ace04c31b8db2d1fc0c18051cd2eefc52a61ea65&vid=4e98bd4c34a9326a56ee3e11797b2e6e1b323c8f&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR