Hydrobromic acid 47%, EMSURE® ACS, ISO for analysis, Supelco®

Supplier: MilliporeSigma
Danger

Synonyms: Hydrogen bromide

1.00307.1000
EM1.00307.1000CS 3862 USD
EM1.00307.1000
Hydrobromic acid 47%, EMSURE® ACS, ISO for analysis, Supelco®
Hydrogen bromide
Formula: HBr
MW: 80.91 g/mol
Density: 1.47 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011323
CAS Number: 10035-10-6
EINECS: 233-113-0
UN: 1788
ADR: 8,II

Order Now

Specification Test Results

Solubility (20 °C) soluble
Melting Point -11 °C
Boiling Point 126 °C (1013 hPa)
Vapor Pressure 10.6 hPa (20 °C)
Density 1.49 g/cm³ (20 °C)
pH (H2O, 20 °C) strongly acid

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR