L(+)-Glutamic acid pH 3.0 - 3.5 (8.6 g/l, H₂O, 25 °C) for biochemistry

Supplier: MilliporeSigma

Synonyms: (S)-(+)-2-Aminopentanedioic acid, L(+)-1-Aminopropane-1,3-dicarboxylic acid, (S)-(+)-α-Glutamic acid, H-Glu-OH, L-Glutamic acid, L(+)-2-Aminopentanedioic acid, (S)-(+)-Glutamic acid, (S)-(+)-1-Aminopropane-1,3-dicarboxylic acid, L(+)-α-Glutamic acid

1.00291.0250
EM1.00291.0250EA 165.49 USD
EM1.00291.0250
L(+)-Glutamic acid pH 3.0 - 3.5 (8.6 g/l, H₂O, 25 °C) for biochemistry
L(+)-Glutamic acid
Formula: HO₂CCH₂CH₂CH(NH₂)CO₂H
MW: 147.13 g/mol
MDL Number: MFCD00002634
CAS Number: 56-86-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR