Formic acid 88-91%, EMSURE® ACS for analysis, Supelco®

Supplier: MilliporeSigma
Danger


M-Clarity™ quality level = MQ300

1.00253.1000
EM1.00253.1000CS 897 USD
EM1.00253.1000
Formic acid 88-91%, EMSURE® ACS for analysis, Supelco®
Formic acid
Formula: HCO₂H
Storage Temperature: Ambient
MDL Number: MFCD00003297
CAS Number: 64-18-6
EINECS: 200-579-1
UN: 1779
ADR: 8,II
Merck Index: 13,04265
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a6cee80f8dc5a4afaf0be47215f1cf258eef7cfd&vid=a77e0bed84455e37172a184511237ad1d178fd11&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay (acidimetric) 89.0 - 91.0 %
Colour ≤ 10 Hazen
Acetic acid (CH₃COOH) ≤ 500 ppm
Chloride (Cl) ≤ 5 ppm
Sulphate (SO₄) ≤ 5 ppm
Sulfite (SO₃) ≤ 10 ppm
Heavy metals (as Pb) ≤ 5 ppm
Ag (Silver) ≤ 0.02 ppm
Al (Aluminium) ≤ 0.05 ppm
Ba (Barium) ≤ 0.05 ppm
Be (Beryllium) ≤ 0.02 ppm
Bi (Bismuth) ≤ 0.10 ppm
Ca (Calcium) ≤ 0.2 ppm
Cd (Cadmium) ≤ 0.05 ppm
Co (Cobalt) ≤ 0.02 ppm
Cr (Chromium) ≤ 0.05 ppm
Cu (Copper) ≤ 0.02 ppm
Fe (Iron) ≤ 2 ppm
Ge (Germanium) ≤ 0.05 ppm
K (Potassium) ≤ 0.1 ppm
Li (Lithium) ≤ 0.02 ppm
Mg (Magnesium) ≤ 0.5 ppm
Mn (Manganese) ≤ 0.05 ppm
Mo (Molybdenum) ≤ 0.02 ppm
Na (Sodium) ≤ 0.5 ppm
NH₄ (Ammonium) ≤ 10 ppm
Ni (Nickel) ≤ 0.05 ppm
Pb (Lead) ≤ 0.02 ppm
Sr (Strontium) ≤ 0.02 ppm
Ti (Titanium) ≤ 0.1 ppm
Tl (Thallium) ≤ 0.05 ppm
V (Vanadium) ≤ 0.05 ppm
Zn (Zinc) ≤ 0.05 ppm
Zr (Zirconium) ≤ 0.1 ppm
Non volatile matter ≤ 10 ppm
Dilution test passes test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR