Chromium(VI) oxide, EMSURE® for analysis, Supelco®

Supplier: MilliporeSigma
Danger

Synonyms: Chromic anhydride, Chromium trioxide

1.00229.0250
EM1.00229.0250CS 2106 USD
EM1.00229.0250
Chromium(VI) oxide, EMSURE® for analysis, Supelco®
Chromium (VI) oxide
Formula: CrO₃
MW: 99.99 g/mol
Melting Pt: 195
MDL Number: MFCD00010952
CAS Number: 1333-82-0
UN: 1463
ADR: 5.1 (6.1, 8) II
Merck Index: 13,02256

Order Now

Specification Test Results

Solubility 1854 g/l (20 °C)
Melting Point 197 °C
Molar Mass 99.99 g/mol
Bulk Density 900 kg/m3
Density 2.7 g/cm3 (20 °C)
pH  - 1 (100 g/l, H2O, 20 °C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR