Orthoboric acid 99.5-100.5% (by alkalimetry), EMSURE® for analysis, Supelco®

Supplier: MilliporeSigma
Danger

Synonyms: Boron trihydroxide, Orthoboric acid, Boron hydroxide, Boric acid, Boracic acid

M-Clarity™ quality level = MQ300

1.00165.9025
EM1.00165.9025EA 1274 USD
EM1.00165.9025
Orthoboric acid 99.5-100.5% (by alkalimetry), EMSURE® for analysis, Supelco®
ortho-Boric acid
Formula: B(OH)₃
MW: 61.83 g/mol
Boiling Pt: 185 °C (1013 hPa)
Melting Pt: 160 °C (dec., lit.)
Density: 1.489 g/cm3 (23 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011337
CAS Number: 10043-35-3
EINECS: 233-139-2
REACH: 01-2119486683-25
Merck Index: 13,01326
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=098dd700154daaf6338ebb270902bb2bc704f27d&vid=6eec7d529090dd4c4b966ed8cd8d057153c5a47b&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR