Benzoic acid for elementary analysis, Supelco®

Supplier: MilliporeSigma
Danger

1.00134.0005
EM1.00134.0005CS 13334 USD
EM1.00134.0005
Benzoic acid for elementary analysis, Supelco®
Benzoic acid
Formula: C₇H₆O₂
MDL Number: MFCD00002398
CAS Number: 65-85-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d561f311b11ea8ab2c8f07253e6748e588eb71aa&vid=dcf5e45072cda5106216b497cd29716270ae772c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Solubility 2.9 g/l (25 °C)
Melting Point 121 - 123 °C
Molar Mass 122.12 g/mol
Bulk Density 500 kg/m3
Boiling Point 249 °C (1013 hPa)
Vapor Pressure 0.001 hPa (20 °C)
Flash Point 121 °C
Density 1.321 g/cm3 (20 °C)
pH 2.5 - 3.5 (H2O, 20 °C) (saturated solution)
Ignition Point 570 °C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR