Acetone ≥99.8% (by GC), EMSURE® Reag. Ph. Eur., ACS, ISO for analysis, Supelco®

Supplier: MilliporeSigma
Danger

Synonyms: 2-Propanone, DMK, Propanone, Dimethyl ketone

1.00014.1000 1.00014.1011 1.00014.2500 1.00014.2511 1.00014.5000 1.00014.6010 1.00014.6190 1.00014.9180 1.00014.4000
EM1.00014.1000CS 517 USD
EM1.00014.1000 EM1.00014.1011 EM1.00014.2500 EM1.00014.2511 EM1.00014.5000 EM1.00014.6010 EM1.00014.6190 EM1.00014.9180 EM1.00014.4000
Acetone ≥99.8% (by GC), EMSURE® Reag. Ph. Eur., ACS, ISO for analysis, Supelco®
Acetone
Formula: CH₃COCH₃
MW: 58.08 g/mol
Boiling Pt: 56.2 (lit.)
Melting Pt: -95.4 (lit.)
Density: 0.791 g/ml (25°C) (lit.)
Flash Pt: <–20 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00008765
CAS Number: 67-64-1
EINECS: 200-662-2
UN: 1090
ADR: 3 II
REACH: 01-2119471330-49
Merck Index: 13,00067
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3de5d8e3bf7a94692d684c8f0ccdfb8500fde76d&vid=24fb2e0ec86974b1803780c7d5b23804a085df64&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Solubility (20 °C) soluble
Melting Point -95.4 °C
Molar Mass 58.08 g/mol
Boiling Point 56.2 °C (1013 hPa)
Vapor Pressure 233 hPa (20 °C)
Refractive Index 1.35868 (20 °C)
Explosion Limit 2.6 - 12.8 %(V)
Density 0.79 g/cm3 (20 °C)
pH 5 - 6 (395 g/l, H2O, 20 °C)
Ignition Point 465 °C DIN 51794
Water absorption 1000 g/kg


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR