Acetonitrile ≥99.5% (by GC), EMSURE® ACS, Reag. Ph. Eur. for analysis, Supelco®

Supplier: MilliporeSigma
Danger

1.00003.1000 1.00003.2500 1.00003.6010 1.00003.6025 1.00003.4000
EM1.00003.1000CS 963 USD
EM1.00003.1000 EM1.00003.2500 EM1.00003.6010 EM1.00003.6025 EM1.00003.4000
Acetonitrile ≥99.5% (by GC), EMSURE® ACS, Reag. Ph. Eur. for analysis, Supelco®
Acetonitrile
Formula: H₃CCN
Boiling Pt: 81…82 °C (lit.)
Melting Pt: –45 °C (lit.)
Density: 0.786 g/ml (25°C) (lit.)
Flash Pt: 2 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00001878
CAS Number: 75-05-8
EINECS: 200-835-2
UN: 1648
ADR: 3,II
REACH: 01-2119471307-38
Merck Index: 13,00071
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cef8be50fcca82050c5bda827ad9c5f57f4c5e27&vid=b80b5c39e654fcf6904b18b58e22a816e8e138b7&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Solubility (20 °C) soluble
Melting Point -45.7 °C
Molar Mass 41.05 g/mol
Boiling Point 81.6 °C (1013 hPa)
Vapor Pressure 97 hPa (20 °C)
Flash Point 2 °C
Refractive Index 1.34
Explosion Limit 3.0 - 17 %(V)
Density 0.786 g/cm3 (20 °C)
Ignition Point 524 °C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR