Acetone:Alcohol (50:50), Supelco®

Supplier: MilliporeSigma

XX0774-76
EM-XX0774-76CS 979 USD
EM-XX0774-76
Acetone:Alcohol (50:50), Supelco®
Acetone:Alcohol

Contains 50% acetone, 45% ethanol, and 5% methanol.

Order Now

Specification Test Results

Color (APHA) 10APHA max
Form Clear liquid
Specific Gravity at 25/25°C 0.7881-0.7901g/cm³

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR