Water, OmniTrace Ultra™ for trace metal analysis, Supelco®
Supplier: MilliporeSigma
M-Clarity™ quality level = MQ100
For low level trace metal analysis. Double-distilled and packaged in ISO Class 5 (FED-STD-209E Class 100/M3.5) cleanroom conditions. Meets ACS specifications for ultratrace purity. Product is double-distilled and packed in class 100 clean room conditions into preleached LDPE bottles. Certificate of Analysis supplied with each shipment.
Formula:
H₂O Boiling Pt: 100 °C (1013 hPa) Melting Pt: 0 °C Density: 1 g/cm³ (4 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00011332 CAS Number: 7732-18-5 EINECS: 231-791-2 Merck Index: 13,10098 |
Specification Test Results
Aluminum (Al) | 20pg/g max |
Antimony (Sb) | 10pg/g max |
Arsenic (As) | 10pg/g max |
Barium (Ba) | 10pg/g max |
Beryllium (Be) | 10pg/g max |
Bismuth (Bi) | 10pg/g max |
Boron (B) | 20pg/g max |
Cadmium (Cd) | 10pg/g max |
Calcium (Ca) | 10pg/g max |
Cerium (Ce) | 10pg/g max |
Cesium (Cs) | 10pg/g max |
Chloride (Cl) | 1ppb max |
Chromium (Cr) | 10pg/g max |
Cobalt (Co) | 10pg/g max |
Color (APHA) | 10 max |
Copper (Cu) | 10pg/g max |
Dysprosium (Dy) | 1pg/g max |
Erbium (Er) | 1pg/g max |
Europium (Eu) | 1pg/g max |
Gadolinium (Gd) | 1pg/g max |
Gallium (Ga) | 10pg/g max |
Germanium (Ge) | 10pg/g max |
Gold (Au) | 10pg/g max |
Hafnium (Hf) | 1pg/g max |
Holmium (Ho) | 1pg/g max |
Indium (In) | 1pg/g max |
Iron (Fe) | 10pg/g max |
Lanthanum (La) | 1pg/g max |
Lead (Pb) | 10pg/g max |
Lithium (Li) | 10pg/g max |
Lutetium (Lu) | 1pg/g max |
Magnesium (Mg) | 10pg/g max |
Manganese (Mn) | 10pg/g max |
Mercury (Hg) | 20pg/g max |
Molybdenum (Mo) | 10pg/g max |
Neodymium (Nd) | 1pg/g max |
Nickel (Ni) | 10pg/g max |
Niobium (Nb) | 10pg/g max |
Palladium (Pd) | 10pg/g max |
Phosphate (PO4) | 1ppb max |
Platinum (Pt) | 10pg/g max |
Potassium (K) | 10pg/g max |
Praseodymium (Pr) | 10pg/g max |
Rhenium (Re) | 10pg/g max |
Rhodium (Rh) | 10pg/g max |
Rubidium (Rb) | 10pg/g max |
Ruthenium (Ru) | 10pg/g max |
Samarium (Sm) | 10pg/g max |
Scandium (Sc) | 10pg/g max |
Selenium (Se) | 50pg/g max |
Silver (Ag) | 10pg/g max |
Sodium (N/A) | 10pg/g max |
Strontium (Sr) | 10pg/g max |
Sulfate (SO4) | 1ppb max |
Tantalum (Ta) | 10pg/g max |
Tellurium (Te) | 1pg/g max |
Terbium (Tb) | 10pg/g max |
Thallium (Tl) | 10pg/g max |
Thorium (Th) | 1pg/g max |
Thulium (Tm) | 10pg/g max |
Tin (Sn) | 10pg/g max |
Titanium (Ti) | 10pg/g max |
Tungsten (W) | 10pg/g max |
Uranium (U) | 1pg/g max |
VaN/Adium (V) | 10pg/g max |
Ytterbium (Yb) | 10pg/g max |
Yttrium (Y) | 1pg/g max |
Zinc (Zn) | 10pg/g max |
Zirconium (Zr) | 10pg/g max |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...