Water, OmniTrace Ultra™ for trace metal analysis, Supelco®

Supplier: MilliporeSigma


M-Clarity™ quality level = MQ100

WX0003-6 WX0003-2
EM-WX0003-6CS 1077 USD
EM-WX0003-6 EM-WX0003-2
Water, OmniTrace Ultra™ for trace metal analysis, Supelco®
Water

For low level trace metal analysis. Double-distilled and packaged in ISO Class 5 (FED-STD-209E Class 100/M3.5) cleanroom conditions. Meets ACS specifications for ultratrace purity. Product is double-distilled and packed in class 100 clean room conditions into preleached LDPE bottles. Certificate of Analysis supplied with each shipment.

Formula: H₂O
Boiling Pt: 100 °C (1013 hPa)
Melting Pt: 0 °C
Density: 1 g/cm³ (4 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011332
CAS Number: 7732-18-5
EINECS: 231-791-2
Merck Index: 13,10098

Order Now

Specification Test Results

Aluminum (Al) 20pg/g max
Antimony (Sb) 10pg/g max
Arsenic (As) 10pg/g max
Barium (Ba) 10pg/g max
Beryllium (Be) 10pg/g max
Bismuth (Bi) 10pg/g max
Boron (B) 20pg/g max
Cadmium (Cd) 10pg/g max
Calcium (Ca) 10pg/g max
Cerium (Ce) 10pg/g max
Cesium (Cs) 10pg/g max
Chloride (Cl) 1ppb max
Chromium (Cr) 10pg/g max
Cobalt (Co) 10pg/g max
Color (APHA) 10 max
Copper (Cu) 10pg/g max
Dysprosium (Dy) 1pg/g max
Erbium (Er) 1pg/g max
Europium (Eu) 1pg/g max
Gadolinium (Gd) 1pg/g max
Gallium (Ga) 10pg/g max
Germanium (Ge) 10pg/g max
Gold (Au) 10pg/g max
Hafnium (Hf) 1pg/g max
Holmium (Ho) 1pg/g max
Indium (In) 1pg/g max
Iron (Fe) 10pg/g max
Lanthanum (La) 1pg/g max
Lead (Pb) 10pg/g max
Lithium (Li) 10pg/g max
Lutetium (Lu) 1pg/g max
Magnesium (Mg) 10pg/g max
Manganese (Mn) 10pg/g max
Mercury (Hg) 20pg/g max
Molybdenum (Mo) 10pg/g max
Neodymium (Nd) 1pg/g max
Nickel (Ni) 10pg/g max
Niobium (Nb) 10pg/g max
Palladium (Pd) 10pg/g max
Phosphate (PO4) 1ppb max
Platinum (Pt) 10pg/g max
Potassium (K) 10pg/g max
Praseodymium (Pr) 10pg/g max
Rhenium (Re) 10pg/g max
Rhodium (Rh) 10pg/g max
Rubidium (Rb) 10pg/g max
Ruthenium (Ru) 10pg/g max
Samarium (Sm) 10pg/g max
Scandium (Sc) 10pg/g max
Selenium (Se) 50pg/g max
Silver (Ag) 10pg/g max
Sodium (N/A) 10pg/g max
Strontium (Sr) 10pg/g max
Sulfate (SO4) 1ppb max
Tantalum (Ta) 10pg/g max
Tellurium (Te) 1pg/g max
Terbium (Tb) 10pg/g max
Thallium (Tl) 10pg/g max
Thorium (Th) 1pg/g max
Thulium (Tm) 10pg/g max
Tin (Sn) 10pg/g max
Titanium (Ti) 10pg/g max
Tungsten (W) 10pg/g max
Uranium (U) 1pg/g max
VaN/Adium (V) 10pg/g max
Ytterbium (Yb) 10pg/g max
Yttrium (Y) 1pg/g max
Zinc (Zn) 10pg/g max
Zirconium (Zr) 10pg/g max


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR