2,2,4-Trimethylpentane ≥95%, Supelco®

Supplier: MilliporeSigma
Danger

Synonyms: Isobutyltrimethylmethane, Isooctane, Isobutyltrimethylethane

TX1395-3
EM-TX1395-3EA 1073 USD
EM-TX1395-3
2,2,4-Trimethylpentane ≥95%, Supelco®
2,2,4-Trimethylpentane

Practical

Formula: (CH₃)₂CHCH₂C(CH₃)₃
MW: 114.23 g/mol
Boiling Pt: 99.2 °C (1013 hPa)
Melting Pt: –107 °C
Density: 0.692 g/cm³ (20 °C)
Flash Pt: –12 °C
Storage Temperature: Ambient
MDL Number: MFCD00008943
CAS Number: 540-84-1
EINECS: 208-759-1
UN: 1262
ADR: 3,II
Merck Index: 13,05212
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=57496edad4db111238ada6124c3cea206ebf33e0&vid=bb9d5eb96909b6bebc6a8709db78c4ea0ef52430&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay (GC) 95% min
Color (APHA) 10 max
Form Clear liquid
Infrared spectrum Conforms to standard

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR