2,2,4-Trimethylpentane ≥99% (by GC), GR ACS, Supelco®

Supplier: MilliporeSigma
Danger

Synonyms: Isobutyltrimethylmethane, Isooctane, Isobutyltrimethylethane

TX1390-30 TX1390-5
EM-TX1390-30EA 4483.16 USD
EM-TX1390-30 EM-TX1390-5
2,2,4-Trimethylpentane ≥99% (by GC), GR ACS, Supelco®
2,2,4-Trimethylpentane

Meets ACS Specifications, Meets Reagent Specifications for testing USP/NF monographs

Formula: (CH₃)₂CHCH₂C(CH₃)₃
MW: 114.23 g/mol
Boiling Pt: 98…99 °C (lit.)
Melting Pt: −107 °C (lit.)
Density: 0.692 g/ml (25°C) (lit.)
Flash Pt: –12 °C
Storage Temperature: Ambient
MDL Number: MFCD00008943
CAS Number: 540-84-1
EINECS: 208-759-1
UN: 1262
ADR: 3,II
Merck Index: 13,05212
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=57496edad4db111238ada6124c3cea206ebf33e0&vid=bb9d5eb96909b6bebc6a8709db78c4ea0ef52430&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay (GC) 99.0% min
Color (APHA) 10 max
Form Clear liquid
Residue after evaporation 0.001% max
Sulfur compounds (as S) 0.005% max
Water-soluble titrable acid 0.0003meq/g max

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR