2,2,4-Trimethylpentane ≥99.5% (by GC), OmniSolv®, Supelco®

Supplier: MilliporeSigma
Danger

TX1389-6 TX1389-1 TX1389-6 TX1389P-1
CATX1389-6CS 3829.55 USD
CATX1389-6 EM-TX1389-1 EM-TX1389-6 EM-TX1389P-1
2,2,4-Trimethylpentane ≥99.5% (by GC), OmniSolv®, Supelco®
2,2,4-Trimethylpentane

Suitable for spectrophotometry, chromatography, and residue analysis. UV cutoff 200 nm. Certificate of analysis on label.

Formula: (CH₃)₂CHCH₂C(CH₃)₃
Boiling Pt: 99.2 °C (1013 hPa)
Melting Pt: –107 °C
Density: 0.692 g/cm³ (20 °C)
Flash Pt: –12 °C
Storage Temperature: Ambient
MDL Number: MFCD00008943
CAS Number: 540-84-1
EINECS: 208-759-1
UN: 1262
ADR: 3,II
Merck Index: 13,05212
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=57496edad4db111238ada6124c3cea206ebf33e0&vid=bb9d5eb96909b6bebc6a8709db78c4ea0ef52430&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay (GC) 99.5% min
Capillary ECD responsive substances (as PCNB) 5ng/L max
Color (APHA) 10 max
Filtered through 0.2 µm filter To pass test
Fluorescence (as quinine base) 250ppt max
Form Clear liquid
Infrared spectrum Conforms to standard
Residue after evaporation 1ppm max
UV Abs. at 200 nm 1.00AU max
UV Abs. at 220 nm 0.05AU max
UV Abs. at 230 nm 0.02AU max
UV Abs. at 250 nm 0.005AU max
UV Abs. at 270 nm 0.005AU max
UV Abs. at 300 nm 0.005AU max
UV Cut-off 200nm max
Water (H2O) 0.01% max


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR