1,2,4-Trichlorobenzene, OmniSolv®, Supelco®

Supplier: MilliporeSigma
Warning

TX1056P-1
EM-TX1056P-1CS 3841 USD
EM-TX1056P-1
1,2,4-Trichlorobenzene, OmniSolv®, Supelco®
1,2,4-Trichlorobenzene
Formula: C₆H₃Cl₃
MW: 181.45 g/mol
Boiling Pt: 214.4
Melting Pt: 17
Density: 1.45 g/cm³ (20 °C)
Flash Pt: 99
Storage Temperature: Ambient
MDL Number: MFCD00000547
CAS Number: 120-82-1
EINECS: 204-428-0
UN: 2321
ADR: 6.1 III
Merck Index: 13,09704

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR