Toluene ≥98% for histology, Supelco®

Supplier: MilliporeSigma
Danger

TX0750-1
EM-TX0750-1EA 746 USD
EM-TX0750-1
Toluene ≥98% for histology, Supelco®
Toluene

Practical

Formula: C₆H₅CH₃
Boiling Pt: 110.6 °C (1013 hPa)
Melting Pt: –95 °C
Density: 0.867 g/cm³ (20 °C)
Flash Pt: 4 °C
Storage Temperature: Ambient
MDL Number: MFCD00008512
CAS Number: 108-88-3
EINECS: 203-625-9
UN: 1294
ADR: 3,II
REACH: 05-2114615130-69
Merck Index: 13,09607
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=44efa31292a7dce6d9e60bcbaec208eb42d34e32&vid=73ada9ad1e894808b6c193cabc4babcfaf320a25&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay (GC) 98% min
Color (APHA) 40 max
Form Clear liquid
Infrared spectrum Conforms to standard


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR