Toluene ≥99.9%, OmniSolv®, Supelco®

Supplier: MilliporeSigma
Danger

TX0737-NP20 TX0737P-1 TX0737-6 TX0737-1
EM-TX0737-NP20EA 1314 USD
EM-TX0737-NP20 EM-TX0737P-1 EM-TX0737-6 EM-TX0737-1
Toluene ≥99.9%, OmniSolv®, Supelco®
Toluene

For spectrophotometry, HPLC, gas chromatography, and residue analysis. Filtered through a 0.2µm filter. UV cutoff 285nm. Lot analysis on label.

Formula: C₆H₅CH₃
Boiling Pt: 110…111 °C (lit.)
Melting Pt: –93 °C (lit.)
Density: 0.865 g/ml (25°C) (lit.)
Flash Pt: 4 °C
Storage Temperature: Ambient
MDL Number: MFCD00008512
CAS Number: 108-88-3
EINECS: 203-625-9
UN: 1294
ADR: 3,II
REACH: 05-2114615130-69
Merck Index: 13,09607
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=44efa31292a7dce6d9e60bcbaec208eb42d34e32&vid=73ada9ad1e894808b6c193cabc4babcfaf320a25&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay (Toluene) 99.9% min
Capillary ECD responsive substances (as PCNB) 5ng/L max
Color (APHA) 10 max
Filtered through 0.2 µm filter To pass test
Form Clear liquid
Infrared spectrum Conforms to standard
Residue after evaporation 3ppm max
UV Abs. at 285 nm 1.00AU max
UV Abs. at 286 nm 0.70AU max
UV Abs. at 288 nm 0.40AU max
UV Abs. at 300 nm 0.10AU max
UV Abs. at 350 nm 0.01AU max
UV Cut-off 285nm max
Water (H2O) 0.02% max


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR