Tetrahydrofuran, OmniSolv®, non-UV, Supelco®

Supplier: MilliporeSigma
Danger

Synonyms: Oxolane, 1,4-epoxy-Butane, Butylene oxide, Butane alpha,delta-oxide, Tetramethylene oxide, THF, Oxacyclopentane, Cyclotetramethylene oxide, Diethylene oxide

TX0282-6 TX0282-1
EM-TX0282-6CS 1059 USD
EM-TX0282-6 EM-TX0282-1
Tetrahydrofuran, OmniSolv®, non-UV, Supelco®
Tetrahydrofuran

Stabilized with 250ppm BHT (butylated hydroxytoluene; 2,6-di-tert-butyl-p-cresol). For chromatography. Can be used for HPLC non-UV applications using refractive index and visible wavelength detectors.

Formula: C₄H₈O
MW: 72.11 g/mol
Boiling Pt: 66 °C (1013 hPa)
Melting Pt: –108.5 °C
Density: 0.888 g/cm³ (20 °C)
Flash Pt: –21.5 °C
Storage Temperature: Ambient
MDL Number: MFCD00005356
CAS Number: 109-99-9
EINECS: 203-726-8
UN: 2056
ADR: 3,II
REACH: 05-2114615188-48
Merck Index: 12,09351
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b2d7611caf0127fbef941994edd1d7e69487d54a&vid=2a64e21bd2e13636c3038adf839b3e9ff18509c5&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay (GC) 99.9% min
Color (APHA) 10 max
Filtered through 0.2 µm filter To pass test
Form Clear liquid
Identity (IR-spectrum) Conforms
Peroxide (as H2O2) 0.01% max
Stabilizer (BHT) 0.0140-0.0310%
Titrable acid 1.0µeq/g max
Water (H2O) 0.03% max

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR