Tetrahydrofuran ≥99.9% (by GC) unstabilized, OmniSolv®, Supelco®

Supplier: MilliporeSigma
Danger

TX0279-6 TX0279-1 TX0279-6 TX0279-7 TX0279P-1
CATX0279-6CS 3931.99 USD
CATX0279-6 EM-TX0279-1 EM-TX0279-6 EM-TX0279-7 EM-TX0279-NP20 EM-TX0279P-1
Tetrahydrofuran ≥99.9% (by GC) unstabilized, OmniSolv®, Supelco®
Tetrahydrofuran

Meets Reagent Specifications for testing USP/NF monographs.

Formula: C₄H₈O
Boiling Pt: 65…67 °C (lit.)
Melting Pt: –108 °C (lit.)
Density: 0.889 g/ml (25°C) (lit.)
Flash Pt: –21.5 °C
Storage Temperature: Ambient
MDL Number: MFCD00005356
CAS Number: 109-99-9
EINECS: 203-726-8
UN: 2056
ADR: 3,II
REACH: 05-2114615188-48
Merck Index: 12,09351
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b2d7611caf0127fbef941994edd1d7e69487d54a&vid=2a64e21bd2e13636c3038adf839b3e9ff18509c5&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Expiration date Exp 12 months from mfg date
Assay (GC) 99.9% min
Color (APHA) 10 max
Filtered through 0.2 µm filter To pass test
Form Clear liquid
Identity (IR-spectrum) Conforms
Peroxide (as H2O2)* 0.02% max
Residue after evaporation 2ppm max
Titrable acid 0.8µeq/g max
UV Abs. at 212 nm 1.00AU max
UV Abs. at 225 nm 0.50AU max
UV Abs. at 250 nm 0.10AU max
UV Abs. at 300 nm 0.005AU max
UV Cut-off 212nm max
Water (H2O) 0.02% max


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR