di-Sodium L(+)-tartrate dihydrate 99.0-101.0%, GR ACS, Supelco®

Supplier: MilliporeSigma

Synonyms: Sodium L-(+)-tartrate dihydrate, L(+)-Tartaric acid disodium salt dihydrate, (R,R)-(+)-Tartaric acid disodium salt dihydrate, (R,R)-(+)-Sodium tartrate dibasic dihydrate, Disodium L-(+)-Tartrate Dihydrate, (R,R)-(+)-Disodium tartrate dihydrate, L(+)-Tartaric acid, disodium salt, dihydrate, Sodium L-tartrate dibasic dihydrate, L(+)-Sodium tartrate dibasic dihydrate, Sodium Tartrate, Dihydrate, (+)-Disodium L-tartrate dihydrate, (+)-Disodium-L-tartrat dihydrate

SX0795-1
EM-SX0795-1CS 1669 USD
EM-SX0795-1
di-Sodium L(+)-tartrate dihydrate 99.0-101.0%, GR ACS, Supelco®
di-Sodium L(+)-tartrate dihydrate

Meets ACS Specifications, Meets Reagent Specifications for testing USP/NF monographs

Formula: NaOOC(CHOH)₂COONa·2H₂O
MW: 230.08 g/mol
Melting Pt: 57 °C
Density: 1.818 g/cm³ (20 °C)
MDL Number: MFCD00150035
CAS Number: 6106-24-7
EINECS: 212-773-3
Merck Index: 12,08833
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=39a75038f4ff5e6c234267b15540823ae361bed4&vid=89289572c48b0c6415f8f11e916a51f66e129474&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay 99.0-101.0%
Ammonium (NH4) 0.003% max
Calcium (Ca) 0.01% max
Chloride (Cl) 5ppm max
Color White or colorless
Form Crystals
Heavy metals (as Pb) 5ppm max
Insoluble matter 0.005% max
Iron (Fe) 0.001% max
Loss on drying at 150°C 15.61-15.71%
pH of a 5% solution at 25°C 7.0-9.0
Phosphate (PO4) 5ppm max
Sulfate (SO4) 0.005% max

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR