Sodium hydroxide 10.0 N in aqueous solution, Supelco®

Supplier: MilliporeSigma
Danger

Synonyms: Caustic soda, Caustic soda, Sodium hydrate, Lye, Soda lye

SX0607N-6
EM-SX0607N-6CS 472 USD
EM-SX0607N-6
Sodium hydroxide 10.0 N in aqueous solution, Supelco®
Sodium hydroxide

NIST Traceable.

Formula: NaOH
Boiling Pt: 119…135 °C (1013 hPa)
Melting Pt: 9…10 °C
Density: 1.53 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003548
CAS Number: 1310-73-2
EINECS: 215-185-5
UN: 1824
ADR: 8,II

Order Now

Specification Test Results

Normality 9.95-10.05N


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR