Sodium carbonate, anhydrous ≥99.5% (dried basis), powder, GR ACS ACS, Supelco®

Supplier: MilliporeSigma
Warning

Synonyms: Soda, Carbonic acid sodium salt, Calcined soda, Disodium carbonate, Carbonic acid disodium salt, Na-X, Bisodium carbonate, Carbonic acid, disodium salt, Soda ash

SX0395-3 SX0395-1 SX0395-20 SX0395-5
EM-SX0395-3CS 1509 USD
EM-SX0395-3 EM-SX0395-1 EM-SX0395-20 EM-SX0395-5
Sodium carbonate, anhydrous ≥99.5% (dried basis), powder, GR ACS ACS, Supelco®
Sodium carbonate

Meets ACS Specifications, Meets Reagent Specifications for testing USP/NF monographs

Formula: Na₂CO₃
MW: 105.99 g/mol
Boiling Pt: 1600
Melting Pt: 854 (lit.)
Storage Temperature: Ambient
MDL Number: MFCD00003494
CAS Number: 497-19-8
EINECS: 207-838-8
REACH: 01-2119485498-19
Merck Index: 14,08596
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9a6ff30f1487cdef86add98b38bfa8eb3dac14f6&vid=c5b129b7acb5c897b9d970ea3130d61fb1934b54&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay (dried basis) 99.5% min
Calcium (Ca) 0.03% max
Chloride (Cl) 0.001% max
Color White
Form Powder
Heavy metals (as Pb) 5ppm max
Insoluble matter 0.01% max
Iron (Fe) 5ppm max
Loss on heating at 285°C 1.0% max
Magnesium (Mg) 0.005% max
Phosphate (PO4) 0.001% max
Potassium (K) 0.005% max
Silica (SiO2) 0.005% max
Sulfur compounds (as SO4) 0.003% max


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR