Sodium bromide ≥99.0% ACS, Supelco®

Supplier: MilliporeSigma

SX0390-1
EM-SX0390-1CS 442 USD
EM-SX0390-1
Sodium bromide ≥99.0% ACS, Supelco®
Sodium bromide

Crystals.

Formula: NaBr
Boiling Pt: 1396 °C (1013 hPa)
Melting Pt: 753 °C
Density: 3.2 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003475
CAS Number: 7647-15-6
EINECS: 231-599-9
Merck Index: 13,08666

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR