Sodium acetate trihydrate 99.0-101.0% (by perchloric acid titration), GR ACS, Supelco®

Supplier: MilliporeSigma

SX0255-5 SX0255-5
TXEMSX02555NYEA 282.63 USD
TXEMSX02555NY EM-SX0255-5
Sodium acetate trihydrate 99.0-101.0% (by perchloric acid titration), GR ACS, Supelco®
Sodium acetate trihydrate

Meets ACS Specifications

Formula: H₃CCOONa·3H₂O
Boiling Pt: 120 - 123 °C (1013 hPa)
Melting Pt: 58 °C
Density: 1.4 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00071557
CAS Number: 6131-90-4
EINECS: 204-823-8
Merck Index: 13,08642
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=313c1d45673fab192565649f8cbb7ddff555de39&vid=1ad37a966605626c95117735c83b16eaea381fbc&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay 99.0-101.0%
Calcium (Ca) 0.001% max
Chloride (Cl) 0.0005% max
Heavy metals (as Pb) 0.0005% max
Insoluble matter 0.005% max
Iron (Fe) 0.0005% max
Magnesium (Mg) 0.0005% max
pH (5%, water) 7.5-9.0%
Phosphate (PO4) 0.0002% max
Potassium (K) 0.005% max
Substances reducing permanganate passes test
Sulfate (SO4) 0.002% max


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR