Silica gel ≥80%, GR ACS 28-200 mesh Grade 12, Supelco®

Supplier: MilliporeSigma

SX0143J-3
EM-SX0143J-3EA 475 USD
EM-SX0143J-3
Silica gel ≥80%, GR ACS 28-200 mesh Grade 12, Supelco®
Silica gel

Meets ACS Specifications

Formula: SiO₂
MW: 60.08 g/mol
Boiling Pt: 2230 °C (1013 hPa)
Melting Pt: 1710 °C
Storage Temperature: Ambient
MDL Number: MFCD00011232
CAS Number: 63231-67-4
REACH: 01-2119379499-16

Order Now

Specification Test Results

Color White
Form Crystals to powder
Mesh size (28-200) 80% min
Suitability for moisture absorption To pass test

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR