Sand, quartz standard, washed, calcinated (Ottawa), Supelco®

Supplier: MilliporeSigma

SX0075-3 SX0075-1 SX0075-30
EM-SX0075-3CS 724 USD
EM-SX0075-3 EM-SX0075-1 EM-SX0075-30
Sand, quartz standard, washed, calcinated (Ottawa), Supelco®
Sand, quartz

Granular, 30 to 40 mesh. Washed and ignited. For boats.

Formula: SiO₂
MW: 60.08 g/mol
Boiling Pt: >2200 °C (1013 hPa)
Melting Pt: 1713 °C
Density: 2.622 to 2.645 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD02100519
CAS Number: 14808-60-7
EINECS: 238-878-4

Order Now

Specification Test Results

Chloride (Cl) 0.015% max
Color Beige
Description To pass test
Form Granules
Soluble substances (in HCl) 0.2% max
Volatile substances 0.2% max

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR