Sand, quartz standard (Ottawa), Supelco®

Supplier: MilliporeSigma

SX0070-3 SX0070-1
EM-SX0070-3CS 531 USD
EM-SX0070-3 EM-SX0070-1
Sand, quartz standard (Ottawa), Supelco®
Sand, quartz
Formula: SiO₂
MW: 60.08 g/mol
Boiling Pt: 2230 °C (1013 hPa)
Melting Pt: 1427 °C
Density: 2.622…2.645 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD02100519
CAS Number: 14808-60-7
EINECS: 238-878-4

Order Now

Specification Test Results

Color Beige
Description To pass test
Form Granules

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR