Formalin 10%, neutral buffered, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger

Synonyms: Paraformaldehyde aqueous solution, Methanal solution, Methylaldehyde solution, Formol, Formaline solution, Formalin, Methylaldehyde

R04586-76 R04586-82
CAR04586-82EA 299.43 USD
CAR04586-82 CAR04586-84 CAR04586-86 CAR04586-76 CAR04586B-82 EM-R04586-76 EM-R04586-86 EM-R04586-90 EM-R04586-82 EM-R04586-84 EM-R04586B-82
Formalin 10%, neutral buffered, Sigma-Aldrich®
Formaldehyde

Formalin 10% W/V, Neutral Buffered

Formula: CH₂O
MW: 30.03 g/mol
MDL Number: MFCD00003274
CAS Number: 50-00-0
UN: 2209
ADR: 8,III

Order Now

Specification Test Results

N/A N/A

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR