Giemsa's stain solution, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger

Synonyms: Azure mixture sicc. Giemsa stain, Azur-Eosin-Methylene blue acc. Giemsa, Giemsa's azur-eosin-methylene blue

R03055-82
CAR03055-74CS 3346.8 USD
CAR03055-74 CAR03055-76 EM-R03055-74 EM-R03055-76 EM-R03055-82
Giemsa's stain solution, Sigma-Aldrich®
Giemsa's stain

Polychromatic stain used primarily to stain blood cells and bone marrow specimens.


Prepared from dyes certified by the Biological Stain Commission (BSC).

Flash Pt: 12 °C
Storage Temperature: Ambient
MDL Number: MFCD00081642
CAS Number: 51811-82-6

Order Now

Specification Test Results

Chemical formula Mixture
Key Applications General Chemistry
Absorbance at maximum (Azure) 0.64-0.77AU
Absorbance maximum wavelength (Azure) 648-652nm
Absorbance maximum wavelength (Eosin) 525-528nm
Performance Passes
Shape of Scan Conforms

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR