Pyridine, anhydrous ≥99.9%, DriSolv®, Supelco®

Supplier: MilliporeSigma
Danger


Packed in septum-sealed bottle.

PX2012-6 PX2012-7 PX2012P-1
EM-PX2012-6CS 1705 USD
EM-PX2012-6 EM-PX2012-7 EM-PX2012P-1
Pyridine, anhydrous ≥99.9%, DriSolv®, Supelco®
Pyridine

Suitable for use in organic synthesis and other anhydrous solvent applications. Clear liquid is filtered through a 0.2µm filter, making it free from particulates. Flammable and toxic.
Elbow adapter with septum cap (EM-692004-2) allows for the removal of solvent under nitrogen atmosphere. Replacement septa (EM-692004S-2), for use with elbow adapter, minimizes air and moisture intrusion once the primary septum is punctured. Drying tube (EM-692010-2) filled with t.h.e.* desiccant indicating silica gel removes solvent under a dry-air atmosphere. Large plug (EM-692012-2) is a replacement plug for bottle top.

Formula: C₅H₅N
Boiling Pt: 115 °C (1013 hPa)
Melting Pt: –44 °C
Density: 0.983 g/cm³ (20 °C)
Flash Pt: 17 °C
Storage Temperature: Ambient
MDL Number: MFCD00011732
CAS Number: 110-86-1
EINECS: 203-809-9
UN: 1282
ADR: 3,II
Merck Index: 13,08060
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3f9c5934940218027dae0d80a1f98b6d5179c0a8&vid=9f2ef46a26e488d366e82869b3acb96652b5dab1&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay (GC) 99.8% min
Appearance Clear liquid, free from particulates
Filtered through 0.2 µm filter To pass test
Residue after evaporation 5ppm max
Water (H2O) 50ppm max


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR