Potassium dihydrogen phosphate ≥99% ACS for HPLC, Supelco®

Supplier: MilliporeSigma

Synonyms: Monopotassiumphosphate, Potassium orthophosphate monobasic, prim.-Potassium phosphate, Potassium dihydrogenorthophosphate, Monopotassium phosphate, Potassium phosphate monobasic, Monobasic Potassium Phosphate, Potassium dihydrogen orthophosphate

PX1562-5
EM-PX1562-5EA 167 USD
EM-PX1562-5
Potassium dihydrogen phosphate ≥99% ACS for HPLC, Supelco®
Potassium dihydrogen phosphate

HPLC Mobile Phase Additive

Formula: KH₂PO₄
MW: 136.09 g/mol
MDL Number: MFCD00011401
CAS Number: 7778-77-0

Order Now

Specification Test Results

Assay 99.0% min
Chloride (Cl) 0.001% max
Form White crystals
Heavy metals (as Pb) 0.001% max
Insoluble matter 0.01% max
Iron (Fe) 0.002% max
Loss on drying (105°C) 0.2% max
Sodium (Na) 0.005% max
Sulfate (SO4) 0.003% max
UV Absorbance (1 M, water, 254 nm) 0.04AU max
pH of a 5% solution at 25°C 4.1-4.5

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR