Potassium fluoride ≥99.0%, GR ACS, Supelco®

Supplier: MilliporeSigma
Danger

PX1467-7
EM-PX1467-7EA 1183 USD
EM-PX1467-7 EM-PX1467-4
Potassium fluoride ≥99.0%, GR ACS, Supelco®
Potassium fluoride

Meets ACS Specifications, Meets Reagent Specifications for testing USP/NF monographs

Formula: KF
MW: 58.1 g/mol
Boiling Pt: 1505 °C (1013 hPa)
Melting Pt: 860 °C
Density: 2.48 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011398
CAS Number: 7789-23-3
EINECS: 232-151-5
UN: 1812
ADR: 6.1,III
Merck Index: 13,07714

Order Now

Specification Test Results

Assay 99.0% min
Chloride (Cl) 0.005% max
Heavy metals (as Pb) 0.001% max
Iron (Fe) 0.001% max
Potassium fluosilicate (K2SiF6) 0.1% max
Sodium (Na) 0.2% max
Sulfate (SO4) 0.005% max
Titrable acid 0.03meq/g max
Titrable base 0.01meq/g max

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR