Potassium acetate ≥99.0%, GR ACS, Supelco®

Supplier: MilliporeSigma

Synonyms: 4-Chloro-N-furfuryl-5-sulfamoylanthranlic acid, Diuretic salt, Acetic acid potassium salt, Acetic acid potassium salt

PX1330-30
EM-PX1330-30EA 3053 USD
EM-PX1330-30 EM-PX1330-1 EM-PX1330-7 EM-PX1330-3
Potassium acetate ≥99.0%, GR ACS, Supelco®
Potassium acetate

Meets ACS Specifications, Meets Reagent Specifications for testing USP/NF monographs

Formula: CH₃COOK
MW: 98.14 g/mol
Melting Pt: 304
Density: 1.57 g/cm³ (25 °C)
Flash Pt: >250 °C
Storage Temperature: Ambient
MDL Number: MFCD00012458
CAS Number: 127-08-2
EINECS: 204-822-2
Merck Index: 13,07687
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=13cc4439b794fe60df11a635ce6d3a03e9609099&vid=06c778a684c28fe03ba0262abdbaa81c094e79a6&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay 99.0% min
Calcium (Ca) 0.005% max
Chloride (Cl) 0.003% max
Color White
Form Crystals or crystalline powder
Heavy metals (as Pb) 5ppm max
Insoluble matter 0.005% max
Iron (Fe) 5ppm max
Magnesium (Mg) 0.002% max
pH of a 5% solution at 25°C 6.5-9.0
Phosphate (PO4) 0.001% max
Sodium (Na) 0.03% max
Sulfate (SO4) 0.002% max


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR