Orthophosphoric acid ≥85.0% for HPLC, Supelco®

Supplier: MilliporeSigma
Danger

PX0996-6 PX0996-6
CAPX0996-6CS 1463.9 USD
CAPX0996-6 EM-PX0996-6
Orthophosphoric acid ≥85.0% for HPLC, Supelco®
Orthophosphoric acid
Formula: H₃PO₄
Melting Pt: 28 °C
Density: 1.71…1.87 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011340
CAS Number: 7664-38-2
EINECS: 231-633-2
UN: 1805
ADR: 8,III

Order Now

Specification Test Results

Max. UV Absorbance at 220 nm 0.05
Max. UV Absorbance at 254 nm 0.04
Max. UV Absorbance at 300 nm 0.02
Antimony 0.002% max.
Arsenic 0.5 ppm max.
Assay 85.0% min.
Color (APHA) 10 max.
Chloride 3 ppm max.
Heavy Metals (as Pb) 0.001% max.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR