Petroleum spirit, 35…60 °C, OmniSolv®, Supelco®

Supplier: MilliporeSigma

Synonyms: Gasoline, Petroleum ether, Petrol, Benzine

PX0424-1 PX0424-1
CAPX0424-1CS 2702.51 USD
CAPX0424-1 EM-PX0424-6 EM-PX0424-1
Petroleum spirit, 35…60 °C, OmniSolv®, Supelco®
Petroleum spirit
Density: 0.7
Flash Pt: -10
MDL Number: MFCD00081849
CAS Number: 64742-49-0

Order Now

Specification Test Results

Boiling range 35-60°C
Capillary ECD responsive substances (as PCNB) 2ng/L max
Color (APHA) 10 max
Filtered through 0.2 µm filter To pass test
Form Clear liquid
Infrared spectrum Conforms to standard
Residue after evaporation 1ppm max
Titrable acid 0.2µeq/g max
Water (H2O) 0.01% max


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR