Ninhydrin, Sigma-Aldrich®

Supplier: MilliporeSigma
Warning

Synonyms: Ninhydrin monohydrate, Trioxohydrindene monohydrate, 2,2-Dihydroxyindane-1,3-dione, 2,2-Dihydroxy-1,3-indanedione, Indanetrione hydrate, 2,2-Dihydroxyindan-1,3-dion, 1,2,3-Indantrione monohydrate

NX0403-2 NX0403-1 NX0403-6
EM-NX0403-2EA 371.06 USD
EM-NX0403-2 EM-NX0403-1 EM-NX0403-6
Ninhydrin, Sigma-Aldrich®
Ninhydrin
Formula: C₉H₆O₄
Boiling Pt: 351 °C (1013 hPa)
Melting Pt: 250…258 °C
Density: 0.862 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003791
CAS Number: 485-47-2
EINECS: 207-618-1
Merck Index: 13,06582
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d0ff1a5cd0c772262dac61a6c63dbdeca97baa2c&vid=61d6c7dbd1fe019587f026a878ce0951ce80fcc8&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay 99% min
Identity (IR-spectrum) Conforms
pH (1%, water) 4.6-5.6
Suitability for detection of amino acids To pass test
Sulfated ash 0.1% max


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR