Molecular sieve A4 (0.4 nm, 4 Å) Grade 514, beads Mesh 8 - 12, Supelco®

Supplier: MilliporeSigma

MX1583L-1
EM-MX1583L-1EA 237 USD
EM-MX1583L-1
Molecular sieve A4 (0.4 nm, 4 Å) Grade 514, beads Mesh 8 - 12, Supelco®
Molecular sieve A4 (0.4 nm, 4 Å)

Beads.

MDL Number: MFCD03457537
CAS Number: 70955-01-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR