Molecular sieve 3A (0.3 nm, 3 Å), beads Mesh 4 - 8, Supelco®
Supplier: MilliporeSigma
Synonyms:
Molecular sieve 0.3 nm, 3A molecular sieve, Molecular sieve A3, Linde 3A, Sodium aluminium silicate
MX1583C-4
EM-MX1583C-4EA
391
USD
EM-MX1583C-4
Molecular sieve 3A (0.3 nm, 3 Å), beads Mesh 4 - 8, Supelco®
Molecular sieve 3A (0.3 nm, 3 Å)
Beads.
Formula:
KnNa₁₂-n[(AlO₂)₁₂(SiO₂)₁₂]·xH₂O |
MDL Number:
MFCD00131613 CAS Number: 308080-99-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...