Molecular sieve 3A (0.3 nm, 3 Å), pellets Mesh 8 - 12, Diameter 1/16", Supelco®

Supplier: MilliporeSigma

Synonyms: Molecular sieve 0.3 nm, 3A molecular sieve, Molecular sieve A3, Linde 3A, Sodium aluminium silicate

MX1583A-1 MX1583A-1
CAMX1583A-1EA 275.35 USD
CAMX1583A-1 EM-MX1583A-1
Molecular sieve 3A (0.3 nm, 3 Å), pellets Mesh 8 - 12, Diameter 1/16", Supelco®
Molecular sieve 3A (0.3 nm, 3 Å)

Pellets.

Formula: KnNa₁₂-n[(AlO₂)₁₂(SiO₂)₁₂]·xH₂O
MDL Number: MFCD00131613
CAS Number: 308080-99-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR