N-Methyl-2-pyrrolidone (NMP), anhydrous ≥99.8%, DriSolv®, Supelco®

Supplier: MilliporeSigma
Danger


Packed in septum-sealed bottle.

MX1396-6
EM-MX1396-6CS 1652 USD
EM-MX1396-6
N-Methyl-2-pyrrolidone (NMP), anhydrous ≥99.8%, DriSolv®, Supelco®
N-Methyl-2-pyrrolidone (NMP)

Clear liquid. Free from particulates. For organic synthesis.

Formula: C₅H₉NO
Boiling Pt: 202 °C (1013 hPa)
Melting Pt: –24 °C
Density: 1.033 g/cm³ (20 °C)
Flash Pt: 91 °C
Storage Temperature: Ambient
MDL Number: MFCD00003193
CAS Number: 872-50-4
EINECS: 212-828-1
Merck Index: 13,06140
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=32d156c242c81ef2974339109ced3895beda87fd&vid=06c87bbb3b5050362f47a8fc836cedc8a6d51530&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay (GC) 99.5% min
Appearance Clear liquid, free from particulates
Color (APHA) 10 max
Residue after ignition 10ppm max
Water (H2O) 50ppm max


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR