N-Methyl-2-pyrrolidone (NMP) for peptide synthesis, Supelco®

Supplier: MilliporeSigma
Danger

MX1394-1
EM-MX1394-1CS 3531 USD
EM-MX1394-1
N-Methyl-2-pyrrolidone (NMP) for peptide synthesis, Supelco®
N-Methyl-2-pyrrolidone (NMP)
Formula: C₅H₉NO
Boiling Pt: 202 °C (1013 hPa)
Melting Pt: –24 °C
Density: 1.033 g/cm³ (20 °C)
Flash Pt: 91 °C
Storage Temperature: Ambient
MDL Number: MFCD00003193
CAS Number: 872-50-4
EINECS: 212-828-1
Merck Index: 13,06140
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=32d156c242c81ef2974339109ced3895beda87fd&vid=06c87bbb3b5050362f47a8fc836cedc8a6d51530&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay (GC) 99.5% min
Appearance Clear, free from particulates
Color (APHA) 20 max
Filtered through 0.2 µm filter To pass test
Form Liquid
Free amine (as dimethylamine) 5ppm max
Heavy metals (as Pb) 1ppm max
Residue after ignition 10ppm max
Titrable base 1.0µeq/g max
UV Abs. at 275 nm 1.00AU max
UV Abs. at 280 nm 0.70AU max
UV Abs. at 300 nm 0.30AU max
UV Abs. at 350 nm 0.10AU max
UV Abs. at 400 nm 0.01AU max
UV Cut-off 275nm max
Water (H2O) 300ppm max


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR