4-Methyl-2-pentanone ≥99.0%, OmniSolv®, Supelco®

Supplier: MilliporeSigma
Danger

MX1301-1
EM-MX1301-1CS 2170 USD
EM-MX1301-1
4-Methyl-2-pentanone ≥99.0%, OmniSolv®, Supelco®
4-Methyl-2-pentanone

For spectrophotometry and chromatography. UV cutoff 334nm. Lot analysis on label.

Formula: (CH₃)₂CHCH₂COCH₃
Boiling Pt: 117.4 °C (1013 hPa)
Melting Pt: –84 °C
Density: 0.801 g/cm³ (20 °C)
Flash Pt: 16 °C
Storage Temperature: Ambient
MDL Number: MFCD00008938
CAS Number: 108-10-1
EINECS: 203-550-1
UN: 1245
ADR: 3,II
Merck Index: 13,05227
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3d44947ef43cbc8103ae523ce5da380ef184ba09&vid=373b9075524004b95d32d3cf9698896714467e8c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay (GC) 99.0% min
Color (APHA) 10 max
Filtered through 0.2 µm filter To pass test
Form Clear liquid
Infrared spectrum Conforms to standard
Residue after evaporation 5ppm max
UV Abs. at 334 nm 1.00AU max
UV Abs. at 339 nm 0.50AU max
UV Abs. at 349 nm 0.20AU max
UV Abs. at 400 nm 0.01AU max
UV Cut-off 334nm max
Water (H2O) 0.08% max


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR