4-Methyl-2-pentanone ≥98.5% (by GC), GR ACS ACS, Supelco®

Supplier: MilliporeSigma
Danger

Synonyms: Methyl iso-Butyl Ketone, Isobutyl Methyl Ketone, Methyl Isobutyl Ketone, MIK

MX1300-30 MX1300-3
EM-MX1300-30EA 4642 USD
EM-MX1300-30 EM-MX1300-3
4-Methyl-2-pentanone ≥98.5% (by GC), GR ACS ACS, Supelco®
4-Methyl-2-pentanone

Meets ACS Specifications, Meets Reagent Specifications for testing USP/NF monographs

Formula: (CH₃)₂CHCH₂COCH₃
MW: 100.16 g/mol
Boiling Pt: 117.4
Melting Pt: -84 (lit.)
Density: 0.8 g/cm3 (20 °C), 0.801 g/ml (25°C) (lit.)
Flash Pt: 16
Storage Temperature: Ambient
MDL Number: MFCD00008938
CAS Number: 108-10-1
EINECS: 203-550-1
UN: 1245
ADR: 3 II
Merck Index: 13,05227
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3d44947ef43cbc8103ae523ce5da380ef184ba09&vid=373b9075524004b95d32d3cf9698896714467e8c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay (GC) 98.5% min
Appearance Clear
Color (APHA) 15 max
Form Liquid
Residue after evaporation 0.005% max
Titrable acid 0.002meq/g max
Water (H2O) 0.1% max


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR