2-Methylbutane ≥95%, Supelco®

Supplier: MilliporeSigma
Danger

Synonyms: iso-Pentane

MX0760-1 MX0760-1
CAMX0760-1EA 635.38 USD
CAMX0760-1 EM-MX0760-1
2-Methylbutane ≥95%, Supelco®
2-Methylbutane

Practical

Formula: CH₃CH₂CH(CH₃)₂
MW: 72.15 g/mol
Boiling Pt: 28 °C (1013 hPa)
Melting Pt: –160 °C
Density: 0.62 g/cm³ (20 °C)
Flash Pt: –51 °C
Storage Temperature: Ambient
MDL Number: MFCD00009338
CAS Number: 78-78-4
EINECS: 201-142-8
UN: 1265
ADR: 3,I
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=498904a9f8b2618e19d2343aa74e247ec7121f00&vid=c7ed2044255d0d72af23a3fafec4ef6066d25097&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay (GC) 95% min
Color (APHA) 30 max
Form Clear liquid
Infrared spectrum Conforms to standard

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR