Ethylene glycol monomethyl ether (2-methoxyethanol) ≥99.3%, GR ACS ACS, Supelco®

Supplier: MilliporeSigma
Danger

Synonyms: Ethylene glycol monomethyl ether

MX0515-6 MX0515-1
EM-MX0515-6CS 1112 USD
EM-MX0515-6 EM-MX0515-1
Ethylene glycol monomethyl ether (2-methoxyethanol) ≥99.3%, GR ACS ACS, Supelco®
Ethylene glycol monomethyl ether (2-Methoxyethanol)

Meets ACS Specifications

Formula: C₃H₈O₂
Boiling Pt: 124.6 °C (1013 hPa)
Melting Pt: –85 °C
Density: 0.964 g/cm³ (20 °C)
Flash Pt: 39 °C
Storage Temperature: Ambient
MDL Number: MFCD00002867
CAS Number: 109-86-4
EINECS: 203-713-7
UN: 1188
ADR: 3,III
Merck Index: 12,06118

Order Now

Specification Test Results

Assay (GC, corrected for water content) 99.3% min
Color (APHA) 10 max
Form Clear liquid
Titrable acid 0.002meq/g max
Water (H2O) 0.1% max


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR