Methanol ≥99.8% (by GC), GR ACS, meets analytical specification for testing USP/NF monographs, Supelco®

Supplier: MilliporeSigma
Danger

MX0485-5 MX0485-3 MX0485-7 MX0485-8 MX0485-9 MX0485-8 MX0485-5 MX0485-6 MX0485-9 MX0485-3 MX0485-20 MX0485-7 MX0485P-4
CAMX0485-5EA 1441.92 USD
CAMX0485-5 CAMX0485-3 CAMX0485-7 CAMX0485-8 CAMX0485-9 EM-MX0485-8 EM-MX0485-5 EM-MX0485-6 EM-MX0485-9 EM-MX0485-3 EM-MX0485-20 EMD-MX0485-7 EMD-MX0485P-4 EMMX0485-31
Methanol ≥99.8% (by GC), GR ACS, meets analytical specification for testing USP/NF monographs, Supelco®
Methanol

Meets ACS Specifications, Meets Reagent Specifications for testing USP/NF monographs.

Formula: CH₃OH
Boiling Pt: 64.7 °C (lit.)
Melting Pt: –98 °C (lit.)
Density: 0.791 g/ml (25°C) (lit.)
Flash Pt: 11 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00004595
CAS Number: 67-56-1
EINECS: 200-659-6
UN: 1230
ADR: 3,6.1,II
REACH: 01-2119433307-44
Merck Index: 13,05984
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5ea983428c5015ad8417c1e1c260cce3a4d1de3c&vid=9bc2f21fdce2dcabce477372374ff56b8fb3a0f1&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Clear
Assay (GC) 99.8% min
Carbonyl (acetone;formaldehyde;acetaldehyde) 0.001% max
Color (APHA) 10 max
Evaporation residue 0.001% max
Solubility in water To pass test
Substances darkened by sulfuric acid To pass test
Substances reducing permanganate To pass test
Titrable acid 0.0003meq/g max
Titrable base 0.0002meq/g max
Water (H2O) 0.1% max


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR