Methanol ≥99.9%, OmniSolv® for purge & trap, Supelco®

Supplier: MilliporeSigma
Danger

Synonyms: Wood alcohol, Methyl alcohol, Methyl hydroxide, Hydroxy methane, Carbinol, Methylol

MX0482-6
EM-MX0482-6CS 1089 USD
EM-MX0482-6
Methanol ≥99.9%, OmniSolv® for purge & trap, Supelco®
Methanol

For use in the preparation of volatile organic standards.

Formula: CH₃OH
MW: 32.04 g/mol
Boiling Pt: 64.6 °C (1013 hPa)
Melting Pt: –98 °C
Density: 0.7918 g/cm³ (20 °C)
Flash Pt: 11 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00004595
CAS Number: 67-56-1
EINECS: 200-659-6
UN: 1230
ADR: 3,II
REACH: 01-2119433307-44
Merck Index: 13,05984
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5ea983428c5015ad8417c1e1c260cce3a4d1de3c&vid=9bc2f21fdce2dcabce477372374ff56b8fb3a0f1&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay (GC) 99.9% min
Color (APHA) 10 max
Form Clear liquid
Titrable acid 0.0003meq/g max
Titrable base 0.0002meq/g max
Volatile impurities (P&T) Methyl ethyl ketone 0.1ppm max
Volatile impurities (P&T) other organics 0.05ppm max

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR