D-(-)-Mannitol, GR ACS, Sigma-Aldrich®

Supplier: MilliporeSigma

Synonyms: Mannitol, (2R,3R,4R,5R)-Hexane-1,2,3,4,5,6-hexaol

MX0214-1
EM-MX0214-1EA 563 USD
EM-MX0214-1
D-(-)-Mannitol, GR ACS, Sigma-Aldrich®
D(-)-Mannitol

Meets ACS Specifications

Formula: HOH₂C(CH(OH))₄CH₂OH
MW: 182.17 g/mol
Boiling Pt: 290…295 °C (4 hPa)
Melting Pt: 164…169 °C
Density: 1.49 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00064287
CAS Number: 69-65-8
EINECS: 200-711-8
Merck Index: 13,05769
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=163f556dca45086e05ff490ddc8eeb65d04aea05&vid=408e3f5566e309064ce68988cdd160305c5b8c0a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Color White
Form Powder
Heavy metals (as Pb) 5ppm max
Insoluble matter 0.01% max
Loss on drying (105°C) 0.05% max
Reducing sugars To pass test
Residue after ignition 0.01% max
Specific rotation [a]25/D +23.3-+24.3°
Titrable acid 0.0008meq/g max

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR